Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Fixable Viability Dye, 750/777 Nm, 50 Tests

Catalog No. 502699019
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-269-9019 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-269-9019 Supplier Abcam Supplier No. AB31468250TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Labeling in less than 4 hours with 30 seconds hands-on time.

  • Assay type: Cell-based
  • Applications: Fluorescence Microscopy, Flow Cyt
  • Sample types: Adherent cells, Suspension cells
  • Results type: Quantitative
  • Reactive species: Mammals
  • Protocol: Labeling in < 4 hrs with 30 secs hands-on time
  • Antibody recovery: 100%

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule Viable Cells
Reactivity Mammals
For Use With (Equipment) Flow Cytometer, Fluorescence Microscope
No. of Reactions 50 Tests
Label or Dye Fixable Viability Dye
Format Kit
Detection Method Fluorescence
Content And Storage Store at -20°C.
Cross Reactivity Mammals
Sample Type Adherent cells, Suspension cells
Quantity 50 Tests
For Use With (Application) Fluorescence Microscopy, Flow Cytometry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.