Learn More
Abcam Fixable Viability Dye, 787/808 Nm, 200 Tests
Description
- Assay type: Cell-based
- Applications: Flow Cyt, Fluorescence Microscopy
- Detection method: Fluorescent
- Sample types: Adherent cells, Suspension cells
- Results type: Quantitative
- Reactive species: Mammals
- Features: Labeling in < 4 hrs with 30 secs hands-on time; 100% antibody recovery
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Reactivity | Mammals |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Label or Dye | Fixable Viability Dye |
| No. of Reactions | 200 Tests |
| For Use With (Equipment) | Flow Cytometer, Fluorescence Microscope |
| Format | Kit |
| Detection Method | Fluorescence |
| Content And Storage | Store at -20°C. |
| Detection Location | In-Cell Detection |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.