Learn More
Sartorius Fixable Viability Kit
Catalog No. 502891622 Shop All Sartorius Products
Encompass
The iQue Fixable Viability Kits are used to label viable from non-viable mammalian cells prior to downstream fixation and permeabilization which is required for intracellular cell staining. The dyes are cell membrane-impermeant that covalently label cell surface and intracellular amines resulting in dim surface staining of live cells and highly fluorescent staining of cells with compromised membranes. The staining is stable and the fluorescence is preserved following fixation and permeabilization.Key AdvantagesPreserve staining post fixation - Maintain fluorescence staining intensity to discriminate live and dead cells post fixation or permeabilization.Enable artifact-free analysis of intracellular targets- Monitor surface and intracellular marker expression simultaneously while reducing artifacts from dead cells.Save precious sample - Conserve samples for use in multiple downstream assays with only 20 L of cells required.
Catalog No. 50-289-1622 Supplier Sartorius Supplier No. BA97122
The iQue Fixable Viability Kits are used to label viable from non-viable mammalian cells prior to downstream fixation and permeabilization which is required for intracellular cell staining. The dyes are cell membrane-impermeant that covalently label cell surface and intracellular amines resulting in dim surface staining of live cells and highly fluorescent staining of cells with compromised membranes. The staining is stable and the fluorescence is preserved following fixation and permeabilization.Key AdvantagesPreserve staining post fixation - Maintain fluorescence staining intensity to discriminate live and dead cells post fixation or permeabilization.Enable artifact-free analysis of intracellular targets- Monitor surface and intracellular marker expression simultaneously while reducing artifacts from dead cells.Save precious sample - Conserve samples for use in multiple downstream assays with only 20 L of cells required.