Learn More
Invitrogen™ eBioscience™ Fixation/Permeabilization Concentrate
Catalog No. 501129082
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 mL
Catalog No. Quantity
50-112-9082 30 mL
1 options
Contains formaldehyde
Catalog No. 50-112-9082 Supplier Invitrogen™ Supplier No. 00512343
Add to Cart
Edge
Add to Cart
Contains formaldehyde
Includes
One 30 mL bottle
Intended to be used with antibodies to transcription factors and nuclear proteins, such as Foxp3 and Ki-67, as well as cytokines and chemokines.
  • This must be used in combination with the Fixation/Permeabilization Diluent (cat. 00-5223) and the Permeabilization Buffer (10X) (cat. 00-8333)
  • Alternatively, all buffers required for staining with these antibodies can be purchased as a set in the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523)
TRUSTED_SUSTAINABILITY
Specifications
Includes One 30 mL bottle
Quantity 30 mL
Reagent Type Fixation Reagent, Permeabilization Reagent
Product Line eBioscience
Product Type Fixation/Permeabilization Buffer
For Research Use Only. Not for use in diagnostic procedures.