Learn More
Invitrogen™ eBioscience™ Fixation/Permeabilization Diluent
Catalog No. 501129081
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
50-112-9081 100 mL
1 options
Contains Sodium azide
Catalog No. 50-112-9081 Supplier Invitrogen™ Supplier No. 00522356
  • $62.50
    $47.65 / Each of 1
    Save $14.85
    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart
Contains Sodium azide
Intended to be used with antibodies to transcription factors and nuclear proteins, such as Foxp3 and Ki-67, as well as cytokines and chemokines.
  • This must be used in combination with the Fixation/Permeabilization Concentrate (cat. 00-5123) and the Permeabilization Buffer (10X) (cat. 00-8333)
  • Alternatively, all buffers required for staining with these antibodies can be purchased as a set in the Foxp3/Transcription Factor Buffer Staining Set (cat. 00-5523)
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mL
Reagent Type Fixation Diluent, Permeabilization Diluent
Product Line eBioscience
Product Type Fixation/Permeabilization Diluent
For Research Use Only. Not for use in diagnostic procedures.