Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Fixation and Permeabilization Flow Cytometry Kit
SDP

Catalog No. NB590630 Shop All Novus Products
Change view
Click to view available options
Quantity:
200 mL (2 x 100 mL)
8 x 5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB590630 8 x 5 mL
NB590373 200 mL (2 x 100 mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB590630 Supplier R&D Systems™ Supplier No. NBP2313778X5ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. For flow cytometric analysis of intracellular antigens

Specifications

Content And Storage Store at room temperature.
For Use With (Application) Flow Cytometry
Quantity 8 x 5 mL
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.