Learn More
BINDER™ Shelf, Reinforced
Shelf, reinforced - positioned at bottom level, max. load 45 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system
$749.00 - $849.21
Specifications
Description | Shelf, reinforced - positioned at bottom level, max. load 45 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system |
---|---|
Material | Steel |
Type | Reinforced Shelf |
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
08-111-300
![]() |
BINDER™
80120287 |
BINDER Chambers KB 53 |
Each for $749.00
|
|
|||||
08-111-301
![]() |
BINDER™
80120288 |
BINDER Chambers KB 115 |
Each for $818.00
|
|
|||||
08-111-299
![]() |
BINDER™
80120817 |
BINDER Chambers KT 115, KT 170 |
Each for $849.21
|
|
|||||
08-111-298
![]() |
BINDER™
80120818 |
BINDER Chambers KT 53 |
Each for $804.80
|
|
|||||
Description
- Reinforced
- Positioned at bottom level
- Maximum load of 45 kg
Specifications
Shelf, reinforced - positioned at bottom level, max. load 45 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system | |
Steel | |
Reinforced Shelf |