Learn More
Medicus Health Fixed Speed Centrifuge, 6 Place
SDP
Supplier:  Medicus Health 5264M1
Encompass
∙ Fixed Speed Centrifuge features an extremely quiet and maintenance free brushless induction motor ∙ Fixed Speed Centrifuge with an RCF of 1350g ∙ Fixed Speed Centrifuge has a 6 Place Rotor for 2-10 mL Tubes ∙ Fixed Speed controller set to 3400 RPM ∙ Timer can run centrifuge for up to 30 minutes with bell signal at end ∙ Lid has a window for speed verification with strobe tachometer ∙ Very quiet running at full speed - only 50 decibels (conversational speech is 60 decibels) ∙ Rotor chamber is accessible for easy cleaning and exterior is smooth for quick cleanup ∙ Shock absorbing suction cup feet help keep centrifuge in place if run out of balance ∙ On/Off switch located on front of unit with "power on" indicator light ∙ Compact Design frees up valuable bench space ∙ Overall Dimensions: 10.2"W x 11.8"D x 7.9"H ∙ Runs on Standard 115V, 60Hz AC; Also available in 220V call for details ∙ One year warranty
Catalog No. NC2033495
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.