Learn More
Drummond™ Fixed-Volume Microdispensers

Description
- Contain borosilicate glass bores with stainless steel plungers, except the 1000μL unit which has precision-molded plastic bores
- All plungers with capacities 25μL and larger have PTFE tips
- The 5μL unit (Cat. No. 21-170-10) will deliver 1 to 5μL
Order Info
Repeat Kit is available for 1000μL unit. Custom spacers are available permitting fractional deliveries on all units; contact your Fisher Scientific™ Customer Service Representative.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Accuracy | ±1% |
| Color | Black |
| Number of Channels | 1 |
| Type | Digital Microdispenser |
| Volume (Metric) | 250 μL |
| For Use With (Application) | Precision molded Plastic bores |
| Includes | 1 plunger, 2 O-rings, 1 collet, 1 coil spring, 3 PTFE tip, 1 spring guide |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.