Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Fixed Volume Micropipette
SCI_ED

Catalog No. S39841 Shop All Eisco Products
Change view
Click to view available options
Volume (Metric):
2 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S39841 2 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S39841 Supplier Eisco™ Supplier No. CH0586B

missing translation for 'validMainframeMsg'

Fixed Volume Micropipette is designed to deliver precise liquid volumes with a high degree of accuracy.

  • Fixed volume micropipette.
  • 2 μL increments.
  • Designed to deliver precise liquid volumes using air displacement technology.
  • 2-stop plunger for sample intake/dispense.
  • Mechanical tip ejector.
  • Ideal for classroom, clinical diagnostic laboratories, and routine tests.

Specifications

Autoclavable Autoclavable
Color Black
Disposable Reusable
For Use With Micropipette Tip
Material Plastic
Model CH0586
Number of Channels 1
Sterility Non-sterile
Type Fix Volume Micropipette
Volume (Metric) 2 μL
For Use With (Application) Clinical Diagnostics, Assays
Length (English) 9 in.
Length (Metric) 22.86 cm
Width (English) 1.5 in.
Width (Metric) 3.81 cm
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.