Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Fixed Volume Micropipettes
SCI_ED

Catalog No. S39845 Shop All Eisco Products
Change view
Click to view available options
Volume (Metric):
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S39845 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S39845 Supplier Eisco™ Supplier No. CH0586F

missing translation for 'validMainframeMsg'

Ideal for clinical diagnostic laboratories and routine tests. Eisco™ Fixed Volume Micropipettes are designed to deliver precise liquid volumes using air displacement technology.

  • Fixed volume micropipette
  • Full size
  • 2 stop plunger for sample intake/dispense
  • Air displacement technology

Specifications

Adjustable Tip Spacing No
Autoclavable Partial
Color Black
Disposable Reusable
For Use With Micropipette Tip
Material Plastic
Model Fixed Volume
Number of Channels 1
Programmability No
Sterility Non-sterile
Type Micropipette
Volume (Metric) 20 μL
For Use With (Application) Clinical Diagnostics, Assays
For Use With (Equipment) Micropipette Tips
Includes Micropipette tip ejector
Warranty 1 Year
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.