Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company™ Fixed Volume Mini-Pipettors Set
SCI_ED

Catalog No. S2335013 Shop All Fischer Technical Company Products
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
Color:
Red
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
S2335013 10 μL White
S2335014 20 μL Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S2335013 Supplier Fischer Technical Company™ Supplier No. 347020SET
Add to Cart
Add to Cart

These fixed volume mini pipettors conveniently come packed with 5 pipettors and a pack of 1000 tips.

This kit comes with five fixed volume mini-pipettors and a pack of 1000 pipette tips.

  • Unique miniature pipettors provide accurate fixed volume dispensing
  • Ideal for micro-volumes and gel loading applications
  • Accepts universal fit, disposable pipette tips

Specifications

Autoclavable Autoclavable
Material Plastic
Sterility Non-sterile
Graduated No
Color White
Volume (Metric) 10 μL
Length (Metric) 9 cm
Type Fixed Volume Pipet
Disposable Reusable
For Use With (Equipment) Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.