Learn More
Medchemexpress LLC (E)-1-[(2R)-2-(2-hydroxyethyl)piperidin-1-yl]-3-(2-phenylpyrazolo[1,5-a]pyridin-3-yl)prop-2-en-1-one | 121524-18-3 | 99.2% | 375.46 | C23H25N3O2 | 10 MG

Supplier: Medchemexpress LLC HY105275A10MG
FK 453 is a potent, selective adenosine A1 receptor antagonist intended for research use. It inhibits human A1 receptor activity with an IC50 of 17.2 nM, appears as an off-white to light yellow solid, and also shows activity at adenosine A2B and in primary rat hepatocytes.
- Potent adenosine A1 receptor antagonist (IC50 17.2 nM).
- Additional activity at adenosine A2B (cAMP accumulation IC50 0.98 μM).
- Inhibits NECA-induced glucose production in primary rat hepatocytes (IC50 8.1 μM).
- High purity: 99.2%.
- Molecular weight: 375.46.
- Off-white to light yellow solid; soluble in DMSO (100 mg/mL).
- Storage recommendations: powder -20°C (3 years); in solvent -80°C (6 months).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.