Learn More
Selleck Chemical LLC Tacrolimus (FK506) 500mg 104987-11-3 FR900506, Fujimycin, Prograf
SDP
Supplier:  Selleck Chemical LLC S5003500MG
Encompass_Preferred
Tacrolimus (FK506, FR900506, Fujimycin, Prograf) is a 23-membered macrolide lactone, it reduces peptidyl-prolyl isomerase activity in T cells by binding to the immunophilin FKBP12 (FK506 binding protein) creating a new complex. Tacrolimus also inhibits the phosphatase activity of calcineurin. Tacrolimus induces vascular endothelial autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-648-5
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.