Learn More
Medchemexpress LLC D-alanine, N-((R)-6-carboxy-N2-(N-(1-oxoheptyl)-D-gamma-glutamyl)-L-lysyl)- | 79335-75-4 | ≥98.0% | 502.57 g/mol | C22H38N4O9 | 5mg
SDP
Supplier:  Medchemexpress LLC HYW7467085MG
Encompass_Preferred
FK-565 is a nucleotide oligomerization domain-1 (NOD1) ligand FK-565 induces murine model with arteritis[1]
Catalog No. 50-002-70267
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.