Learn More
Enzo Life Sciences FK-866 (5mg). CAS: 658084-64-1
SDP
Supplier:  Enzo Life Sciences ALX270501M005
Encompass_Preferred
Selective inhibitor of the nicotinamide pathway dependent NAD+synthesis, causing NAD+ depletion. Highly specific, non-competitive inhibitor of nicotinamide phosphoribosyltransferase (NAMPT/NAPRT) for both the enzyme/substrate complex and the free enzyme (Ki=0.4 nM and Ki´=0.3 nM, respectively). NAD+ depletion by FK-866 directs delayed cell death by apoptosis in Hep-G2 human liver carcinoma cells (IC50=~1 nM). Causes premature senescence in normal human smooth muscle cells. Induces autophagy in SH-SY5Y neuroblastoma cells, as indicated by the formation of LC3-positive vesicles. Alternative name: K 22.175, N-[4-(1-benzoyl-4-piperidinyl)butyl]-3-(3-pyridinyl)-2E-propenamide. Purity: ≥98% (NMR). Solubility: Soluble in 100% ethanol, dimethyl formamide (40mg/ml) or DMSO (25mg/ml); sparingly soluble in aqueous buffers. Long Term Storage: -20°C.
Catalog No. 50-200-8624
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.