Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FK506 binding protein associated protein Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987678
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7678 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7678 Supplier Bioss Supplier No. BS7030R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a phosphorylated protein that is a member of a Skp1-Cullin-F-box-like complex. The protein is essential for normal development of the vasculature and mutations in this gene have been associated with glomuvenous malformations, also called glomangiomas. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.

Specifications

Antigen FK506 binding protein associated protein
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GLMN
Gene Accession No. Q92990
Gene Alias 9330160J16Rik; AW227515; BOS_3276; FAP; Fap48; FAP68; FK506-binding protein-associated protein; FKBP associated protein; FKBPAP; FKBP-associated protein; GLML; GLMN; Glomulin; glomulin, FKBP associated protein; GVM; VMGLOM
Gene Symbols GLMN
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FK506 binding protein associated protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11146
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.