Learn More
Apexbio Technology LLC highly specific non-competitive NAMPT inhibitor 658084-64-1. MFCD10565943
SDP
Supplier:  Apexbio Technology LLC A43815
Encompass_Preferred
FK866 is an inhibitor of nicotinamide phosphoribosyltransferase (NMPRTase) with IC50 values ranging between 0.09 nM and 27.2 nM. NAD plays a vital role in numerous biochemical and biologic processes. Targeting NAD synthesis is thought to be a selective manner to kill cancer cells since cancer cells have a higher rate of NAD turnover compared with normal cells. In the in vitro MTT assay using a panel of 41 human hematologic cancer. Other sizes are available. Please inqury us for quote.
Catalog No. 50-101-3518
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.