Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKBP12 Recombinant Rabbit Monoclonal Antibody (1H38L2)

Catalog No. PI702243 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI702243 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI702243 Supplier Invitrogen™ Supplier No. 702243
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Pig, Cat Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

FKBP12 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. The protein is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin. It interacts with several intracellular signal transduction proteins including type I TGF-beta receptor. It also interacts with multiple intracellular calcium release channels, and coordinates multi-protein complex formation of the tetrameric skeletal muscle ryanodine receptor. In mouse, deletion of this homologous gene causes congenital heart disorder known as noncompaction of left ventricular myocardium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP12
Applications Western Blot
Classification Recombinant Monoclonal
Clone 1H38L2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene FKBP1A
Gene Accession No. P26883, P62942, Q62658
Gene Alias 12 kDa FK506-binding protein; 12 kDa FKBP; calstabin 1; calstabin-1; EC 5.2.1.8; FK506 binding protein 1a; FK506 binding protein 1A, 12kDa; FK506 binding protein 2 (13 kDa); FK506 binding protein12; FK506-binding protein 1; FK506-binding protein 1 (12kD); FK506-binding protein 12; FK506-binding protein 1a; FK506-binding protein 1A (12kD); FK506-binding protein, T-cell, 12-kD; Fkbp; FKBP1; FKBP12; FKBP-12; FKBP12-Exip3; Fkbp1a; FKBP-1A; Fkbp2; Immunophilin FKBP12; peptidyl-prolyl cis-trans isomerase FKBP1A; PKC12; PKCI2; PPIASE; PPIase FKBP1A; protein kinase C inhibitor 2; rotamase
Gene Symbols FKBP1A
Host Species Rabbit
Immunogen Peptides corresponding to human FKBP12 [1) aa72-85 2) aa55-66].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100763474, 14225, 2280, 25639
Target Species Hamster, Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.