Learn More
Description
PA1-020 detects FK506 binding protein 51 kDa (FKBP51) in canine, hamster, human, mouse and rat tissues. PA1-020 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~51 kDa protein representing FKBP51 in HeLa cell lysate. By Western blot PA1-020 also detects FKBP51 in whole tissue extracts from rat kidney and rat and mouse testes. The PA1-020 immunogen is a synthetic peptide corresponding to the residues C(394) Q K K A K E H N E R D R R(407) of human FKBP51. There is a single amino acid substitution (K396 to R396) in this domain between human and mouse FKBP51 protein. PA1-020 immunizing peptide (Cat. # PEP-078) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | FKBP5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | FKBP5 |
| Gene Accession No. | Q13451, Q64378 |
| Gene Alias | 51 kDa FK506-binding protein; 51 kDa FKBP; 54 kDa progesterone receptor-associated immunophilin; AIG6; androgen-regulated protein 6; D17Ertd592e; Dit1; EC 5.2.1.8; FF1 antigen; FK506 binding protein 5; FK506-binding protein 5; FKBP prolyl isomerase 5; Fkbp5; FKBP-5; FKBP51; FKBP-51; FKBP54; HSP90-binding immunophilin; p54; peptidylprolyl cis-trans isomerase; Peptidyl-prolyl cis-trans isomerase FKBP5; PPIase; PPIase FKBP5; Ptg-10; rotamase; T-cell FK506-binding protein |
| Gene Symbols | FKBP5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
