Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP8 Mouse anti-Human, Mouse, Clone: 1C1G12, Proteintech
SDP

Catalog No. 501737107 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7107 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7107 Supplier Proteintech Group Inc Supplier No. 666901IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FKBP8 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, it does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function.The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, this encoded protein does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function.

Specifications

Antigen FKBP8
Applications Western Blot
Classification Monoclonal
Clone 1C1G12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene FKBP8
Gene Accession No. O35465, Q14318
Gene Alias 38 kDa FK506 binding protein, 38 kDa FKBP, FK506 binding protein 8, FK506 binding protein 8, 38kDa, FKBP 38, FKBP 8, FKBP38, FKBP8, FKBPr38, hFKBP38, PPIase FKBP8, Rotamase
Gene Symbols Fkbp8
Host Species Mouse
Immunogen FKBP8 Fusion Protein Ag1649
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14232, 23770
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.