Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FKBP8 Recombinant Protein

Catalog No. 89012402 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-012-402 10 μg
89-012-403 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-012-402 Supplier Abnova™ Supplier No. H00023770P0110
Add to Cart
Add to Cart

Human FKBP8 full-length ORF ( AAH09966, 1 a.a. - 355 a.a.) recombinant protein with GST-tag at N-terminal

The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, this encoded protein does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function. (provided by RefSeq)

  • Molecular weight: 64.79kDa
  • Preparation method: in vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 Fast Flow
  • Storage Buffer:50mM Tris-HCI, 10mM reduced Glutathione, pH 8 in the elution buffer
  • Quality Control Testing: 12.5% SDS-PAGE stained with Coomassie Blue

Best use within three months from the date of receipt of this protein

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH09966
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 23770
Molecular Weight (g/mol) 64.79
Name FKBP8 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MGQPPAEEAEQPGALAREFLAAMEPEPAPAPAPEEWLDILGNGLLRKKTLVPGPPGSSRPVKGPVVTVHLQTSLENGTRVQEEPELVFTLGDCDVIQALDLSVPLMDVGETAMVTADSKYCYGPQGRSPYIPPHAALCLEVTLKTAVDGPDLEMLTGQERVALANRRRECGNAHYQRADFVLAANSYDLAIKAITSSAKVDMTFEEEAQLLQLKVKCLNNLAASQLKLDHYRAALRSCSLVLEHQPDNIKALFRKGKVLAQQGEYSEAIPILRAALKLEPSNKTIHAELSKLVKKHAAQRSTETALYRKMLGNPSRLPAKCPGKGAWSIPWKWLFGATAVALGGVALSVVIAARN
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias FKBP38/FKBPr38
Common Name FKBP8
Gene Symbol FKBP8
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.