Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7209088 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1971 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1971 Supplier Proteintech Group Inc Supplier No. 180731AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FKBP9 is a PPIase which accelerate the folding of proteins during protein synthesis.

Specifications

Antigen FKBP9
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene FKBP9
Gene Accession No. O95302, Q66H94, Q9Z247
Gene Alias 63 kDa FK506 binding protein, 63 kDa FKBP, FK506 binding protein 9, FKBP 63, FKBP 9, FKBP60, FKBP63, FKBP9, PPIase, PPIase FKBP9, Rotamase
Gene Symbols FKBP9
Host Species Rabbit
Immunogen FKBP9 Fusion Protein Ag12715
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11328, 27055, 297123
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.