Learn More
Apexbio Technology LLC FLAG tag Peptide, 98849-88-8, MFCD02262135, 4 mg
SDP
Supplier:  Apexbio Technology LLC A60024
Encompass_Preferred
MF: C41H60N10O20, MW: 1012.97, Solubility: 50.6 mg/mL in DMSO, 210.6 mg/mL in H2O, 34.03 mg/mL in EtOH. A fusion tag called FLAG and consisting of eight amino acids Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys including an enterokinase-cleavage site, was specifically designed for immunoaffinity chromatography.
Catalog No. 50-101-2231
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.