Learn More
Enzo Life Sciences Flagellin (high purity) (10 µg)
SDP
Supplier:  Enzo Life Sciences ALX522058C010
Encompass_Preferred
Concentration: 0.1mg/ml after reconstitution. Endotoxin content: Salmonella typhimurium strain 14028. Specificity: Binds to human and mouse TLR5 (Toll-like receptor 5). Use/Stability: Stable for at least 6 months after receipt when stored at -20°C. UniProt: P06179. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -20°C. Long Term Storage: -20°C
Catalog No. NC9041128
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.