Learn More
Description
Specifications
Specifications
| Accession Number | P06179 |
| Concentration | LYOPH |
| For Use With (Application) | Bioactivity, ELISA |
| Formulation | Lyophilized from PBS |
| Quantity | 100 μg |
| Storage Requirements | Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | <0.1 EU/μg purified protein (LAL test; Lonza). |
| Reconstitution | Reconstitute with 100 ul sterile water (1mg/ml after reconstitution) |
| Research Category | Immunology, Innate Immunity |
| Product Line | Novus |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
