Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CritiCore Protective Wear Flame-Resistant Indura Lab Coat

Catalog No. 18111240
Change view
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-111-240 3X-Large
18-111-233 2X-Small
18-111-234 X-Small
18-111-235 Small
18-111-236 Medium
18-111-237 Large
18-111-238 X-Large
18-111-239 2X-Large
18-111-241 4X-Large
18-111-242 5X-Large
18-111-243 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 18-111-240 Supplier CritiCore Protective Wear Supplier No. 12L34940IN303P3XLBLU
Add to Cart
Add to Cart
This item is not returnable. View return policy

Flame-Resistant Indura Lab Coats are durable enough to withstand industrial laundering while maintaining fire-resistant properties that are guaranteed for the life of the garment.

  • 360° fire-resistant identification patches
  • Quick-release snap front
  • Safety-sleeve cuff with tunneled elastic
  • Breast pocket with pencil slot
  • 2 oversized hip pockets
  • Vent openings on the sides allow access to pants pockets
  • Meets HRC1 and NFPA 70E requirements
  • Fire-resistant properties guaranteed for life of garment

Specifications

Size 3X-Large
Gender Unisex
Material FR Cotton
Color Blue
Closure Type Snaps
Cuff Style Tunnelized Elastic
Disposable Reusable
Sterility Non-sterile
No. of Pockets 3
Collar Type Lapel
Hazard Rating HRC 1 Flame-resistant compliant
Certifications/Compliance HRC 1 and NFPA 70E compliant
For Use With (Application) Research Labs, Pharmaceutical, Bio-Tech, Chemistry
Length (English) 39 in.
Length (English) Sleeve 26.5 in.
Length (Metric) 99.06 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.