Learn More
CritiCore Protective Wear Flame-Resistant Women's Lab Coat
Catalog No. 18111334
Change view
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-111-334 3X-Large
18-111-327 2X-Small
18-111-328 X-Small
18-111-329 Small
18-111-330 Medium
18-111-331 Large
18-111-332 X-Large
18-111-333 2X-Large
18-111-335 4X-Large
18-111-336 5X-Large
18-111-337 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 18-111-334 Supplier CritiCore Protective Wear Supplier No. 12L34950IN303P3XLBLU
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
These flame-resistant lab coats were developed specifically to fit the female form while also providing the protection required in many of today’s high-tech laboratories and research facilities.
  • Best in class flammability protection
  • Meets NFPA 70E and HRC 1 performance standards
  • Reusable Indura™ FR fabric
  • Quick-release snap front
  • Safety sleeve cuff with tunneled elastic
  • Princess-belt on back
  • FR identification patches on sleeves and back
  • Excellent heat-resistance properties
Specifications
Size 3X-Large
Gender Unisex
Material FR Cotton
Color Blue
Closure Type Snaps
Cuff Style Elastic
Disposable Reusable
Sterility Non-sterile
No. of Pockets 3
Collar Type Lapel
Latex Free Yes
Arc Flash Rating HRC 1
Certifications/Compliance NFPA 70 E compliant