Learn More
New Pig Flammable Storage Safety Cabinet - Manual Closing
SDP
Catalog No. 17987826 Shop All New Pig Corporation Products
Catalog No. 17-987-826 Supplier New Pig Supplier No. CAB924YW
Add to Cart
Add to Cart
Store flammable materials in this large flammable-safety cabinet, built to be FM-Approved and to meet OSHA and NFPA specifications
  • Securely store flammable materials: Designed to contain hazardous liquids and prevent them from spreading in the event of a fire.
  • Manual-closing door: The door must be manually shut after each use, providing a simple and reliable closure mechanism.
  • Meets safety standards: This cabinet is built to be FM-Approved and to meet OSHA and NFPA specifications, ensuring compliance and a high level of safety.
  • Durable, fire-resistant construction: Features an 18-gauge double-walled steel body with insulating air space to withstand fire and protect contents.
  • Leakproof sump: A built-in sump at the bottom of the cabinet safely contains any drips or spills, preventing them from reaching the floor.
  • Built-in lock: The cabinet is equipped with a non-sparking, three-point latch and a built-in lock with keys, preventing unauthorized access
Specifications
Door Style Manual-close, Locking, Double
Material Galvanized Steel