Learn More
Marketlab Flanged Tube Caps
Description
- Designed with two flexible flanges to provide a leak-proof seal
- Polyethylene construction
- Grooved top eases grip for easy on/off
Specifications
Specifications
| Product Type | Test Tube Plug Cap |
| Autoclavable | Not Autoclavable |
| Material | Polyethylene |
| Color | White |
| Sterility | Non-sterile |
| Disposable | Single-use |