Learn More
MilliporeSigma™ Supelco™ Flangeless Fittings Kit (Upchurch)
Catalog No. 111004033 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Flangeless Fittings Kit (Upchurch)
Catalog No.
11-100-4033 Flangeless Fittings Kit (Upchurch)
1 options
Catalog No. 11-100-4033 Supplier MilliporeSigma™ Supelco™ Supplier No. 58630
Add to Cart
Add to Cart
All fittings are 1/4-28 unless otherwise indicated.
Kit Components:
  • Flangeless ferrules, 1/16 in. (10 ea.)
  • Flangeless ferrules, 1/8 in. (10 ea.)
  • Flangeless nuts, 1/16 in. (10 ea.)
  • Flangeless nuts, 1/8 in. (10 ea.)
  • Unions with fittings, 1/16 in. (2 ea.)
  • Unions with fittings, 1/8 in. (2 ea.)
  • Tee with fittings, 1/16 in. (1 ea.)
  • Tee with fittings, 1/8 in. (1 ea.)
  • Cross with fittings, 1/16 in. (1 ea.)
  • Cross with fittings, 1/8 in. (1 ea.)
  • Unions, metric M6 (2 ea.)
  • Unions, standard 1/4-28 (2 ea.)
  • Luer fittings, female (2 ea.)
  • Luer fittings, male (2 ea.)
  • Metric adapters, 1/4-28 to M6 (2 ea.)
TRUSTED_SUSTAINABILITY
Specifications
Type Flangeless Fittings Kit
For Use With (Application) Chromatography Applications