Learn More
Description
- Store vial at ≤ -20 °C
- 100X FLARE™ BSA Additive is a component of the Recombinant E. coli Fpg FLARE™ Assay Kit, Recombinant E. coli Fpg FLARE™ Module, Recombinant Human OGG1 FLARE™ Assay Kit, and Recombinant Human OGG1 FLARE™ Module
Specifications
Specifications
| Content And Storage | Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| For Use With (Equipment) | Recombinant E. coli Fpg FLARE™ Assay Kit, Recombinant E. coli Fpg FLARE™ Module, Recombinant Human OGG1 FLARE™ Assay Kit, and Recombinant Human OGG1 FLARE™ Module |
| Quantity | 100 μL |
| Species | Bovine |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
