Learn More
Specifications
Specifications
| Content And Storage | Store the unopened product at 2 - 8°C. Do not use past expiration date. |
| For Use With (Application) | Detect specific DNA damage in single cells using DNA repair enzymes in conjunction with Trevigen’s CometAssay™ single cell gel electrophoresis kits |
| Includes | Lysis Solution (2 x 500 mL, store at room temp.), Comet LMAgarose (15 mL, store at 4°C), and 25 FLARE™ Slides |
| Quantity | 75 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
