Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ FLARE™ Slides

Catalog No. 395007502
Change view
Click to view available options
Quantity:
100 Slides
25 Slides
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-500-7502 25 Slides
39-503-0002 100 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-500-7502 Supplier R&D Systems™ Supplier No. 395007502
Add to Cart
Add to Cart
This item is not returnable. View return policy

A component of the FLARE™ Assay Kit, which is used for the rapid analysis of DNA adducts revealed using the CometAssay in conjunction with DNA repair enzymes

  • The CometAssay, or single cell gel electrophoresis assay, permits the quantification of DNA damage in individual cells based on the migration of DNA structures in an electric field
  • As a result of the migration, Comet tail shaped images are formed and the amount of DNA in the Comet tail as well as the distance of migration, can be a measure of damage
  • FLARE™ Slides are convenient, simple, and reliable format for sample preparation, processing, analysis, and storage for the CometAssay and FLARE™ Assay

Specifications

Content And Storage Store the unopened product at room temperature. Do not use past expiration date.
Description Sample-ready pre-treated glass slides
Format Hydrophobic Barrier, Specially-treated Surface, Writing Surface
For Use With (Equipment) For use with CometAssay™ Kit and FLARE™ Assay Kits, reagents, and protocols
For Use With (Application) Sample preparation, processing, analysis, and storage for the CometAssay™ and FLARE™ Assay
Quantity 25 Slides
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.