Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Flash Photolysis Spectrometer
SDPSCI_ED

Catalog No. s40810 Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Flash Photolysis Spectrometer software
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
S40810 Flash Photolysis Spectrometer software
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S40810 Supplier Vernier Supplier No. VSPFP
Only null left
Add to Cart
Add to Cart
Request a Quote

The Vernier Flash Photolysis Spectrometer is a simple, user-friendly device for demonstrating the fundamental principles of chemical kinetics and photochemistry to undergraduate chemistry students. It measures the absorption and emission changes over time of a photoexcited sample at a single wavelength, which is determined by a filter (600 nm filter included).

Photochemical reactions are very important in many areas of chemistry with applications including photodynamic therapy and solar energy harvesting. These reactions often proceed quickly and, as a result, require fast reaction techniques to analyze. The Vernier Flash Photolysis Spectrometer is a simple, user-friendly transient absorption spectrometer. It is useful for demonstrating the fundamental principles of these types of fast, photo-catalyzed reactions. Some experiment ideas include kinetics of photochemical reactions such as

  • Excited-state dynamics
  • Triplet-decay analysis
  • Time-resolved phosphorescence
  • Phosphorescence-quenching kinetics
  • Isomerization
  • Reactive free radicals
  • Photocatalysis

Specifications

Includes Vernier Flash Photolysis Spectrometer; AC power supply; One fluorescence quartz cuvette; USB cable; Empty filter housing (fits standard size, third party filters: 12.5 mm OD and 9.0 mm ID); 600 nm filter (10 nm FWHM) in filter housing; Quick start guide
Grade Recommended for College
For Use With (Equipment) Flash Photolysis Spectrometer software
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.