Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FLASH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA146274
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA146274 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA146274 Supplier Invitrogen™ Supplier No. PA146274
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FLASH Polyclonal Antibody for Western Blot

This protein is highly similar to FLASH, a mouse apoptotic protein identified by its interaction with the death-effector domain (DED) of caspase 8. Studies of FLASH protein suggested that this protein may be a component of the death-inducing signaling complex that includes Fas receptor, Fas-binding adapter FADD, and caspase 8, and plays a regulatory role in Fas-mediated apoptosis. Alternative splicing results in multiple transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FLASH
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CASP8AP2
Gene Accession No. Q9UKL3, Q9WUF3
Gene Alias AA387232; CASP8 associated protein 2; CASP8AP2; CASP8-associated protein 2; caspase 8 associated protein 2; CED-4; D4Ertd659e; FLASH; FLASH homolog RIP25; FLICE associated huge; FLICE-associated huge protein; FLJ11208; human FLASH; KIAA1315; mKIAA1315; RIP25; RP11-63K6.3
Gene Symbols CASP8AP2
Host Species Rabbit
Immunogen C-terminus of the FLASH protein (proprietary peptide).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26885, 9994
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.