Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azure Biosystems Flash Western Chemiluminescence Kit
SDP

Catalog No. p-200054226 Shop All Azure Biosystems Products
Change view
Click to view available options
Conjugate:
Flash Anti-mouse HRP Conjugate
Flash Anti-rabbit HRP Conjugate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
AC2221AZ Flash Anti-mouse HRP Conjugate
AC2220AZ Flash Anti-rabbit HRP Conjugate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC2221AZ Supplier Azure Biosystems Supplier No. AC2221
Only null left
Add to Cart
Add to Cart

Kit includes all reagents necessary for performing 10 chemiluminescent Western blots using the Flash Western protocol. Contains LF PVDF membranes, Azure Blocking Buffer, Conjugate, Flash Wash Buffer and Radiance ECL substrate.

  • 9 x 7 cm Low Fluorescence PVDF Membranes
  • 10 membranes
  • Either Anti-mouse or Anti-Rabbit HRP Conjugate

Specifications

Product Type Western Blot Kit
Detection Method Chemiluminescence
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Quantity 10 Mini Blots
Conjugate Flash Anti-mouse HRP Conjugate
Substrate Horseradish Peroxidase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.