Learn More
Description
FlashGel™ FLX DNA Prestain Loading Buffer is a ready to use sample buffer combined with a DNA specific dye, formulated for preparing and staining DNA samples prior to loading onto stain free FlashGel™ FLX Cassettes. Simply mix your DNA samples with the buffer, load them into the cassette, and track band migration in real time. Results are generated in under 5 minutes, enabling fast DNA analysis directly at the bench. Together, FlashGel™ FLX Cassettes and the DNA Prestain Loading Buffer provide a streamlined, flexible workflow for rapid separation and easy documentation. Compatiblity: FlashGel™ FLX Cassettes used with FlashGel™ FLX DNA Prestain Loading Buffer are fully compatible with the existing FlashGel™ System components, including FlashGel™ Dock, Camera, and Markers.
- Optimized for DNA separation using FlashGel™ FLX Cassettes
- Long shelf life supports reliable storage and routine use
- High sensitivity with sharp, clear band resolution
- Real-time separation with instant documentation
- Capture images directly at the bench no UV transilluminator required
Specifications
Specifications
| Content And Storage | Contains 1 x 1 mL 5X FlashGel FLX DNA Loading Buffer. Store buffer at room temperature. Buffer has a minimum shelf life of 6 months from the date of shipping. |
| Buffer | FLX DNA Loading Buffer |
| Concentration | 5X |
| Format | Tube |
| Gel Compatibility | FlashGel FLX Cassettes |
| Quantity | 1 x 1 mL |
| Shipping Condition | Room Temperature |
| Product Type | DNA Sample Loading Buffer |
| For Use With (Equipment) | FlashGel System |
For research use only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.