Learn More
Description
FlashGel™ FLX RNA Prestain Loading Buffer is a ready to use sample buffer combined with a RNA specific dye, formulated for preparing and staining RNA samples prior to loading onto stain free FlashGel™ FLX Cassettes. Simply mix your RNA samples with the buffer, load them into the cassette, and track band migration in real time. Results are generated in under 5 minutes, enabling fast RNA analysis directly at the bench. Together, FlashGel™ FLX Cassettes and the RNA Prestain Loading Buffer provide a streamlined, flexible workflow for rapid separation and easy documentation. FlashGel™ FLX RNA Prestain Loading Buffer supports the preparation of both native and denatured RNA samples. For denatured RNA, samples must be heat-treated to ensure effective separation. Compatiblity: FlashGel™ FLX Cassettes used with FlashGel™ FLX RNA Prestain Loading Buffer are fully compatible with the existing FlashGel™ System components, including FlashGel™ Dock, Camera, and Markers. Main benefits are:
- Optimized for RNA separation using FlashGel™ FLX Cassettes
- Long shelf life supports reliable storage and routine use
- Detect <10 ng RNA per band
- Real‐time separation with instant documentation
- Capture images directly at the bench no UV transilluminator required
- Avoid hazardous reagents and contaminating RNases
Specifications
Specifications
| Content And Storage | Contains 1 x 1 mL 2X FlashGel FLX RNA Loading Buffer. Store buffer at -20°C. Buffer has a minimum shelf life of 6 months from the date of shipping. |
| Buffer | FLX RNA Loading Buffer |
| Color | Blue |
| Concentration | 2X |
| Format | Tube |
| Gel Compatibility | FlashGel FLX Cassettes |
| Quantity | 1 X 1 mL |
| RNase Free | 24 months from date of manufacture |
| Product Type | RNA Sample Loading Buffer |
| For Use With (Equipment) | FlashGel System |
For research use only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.