Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ FlashGel™ System DNA Cassettes

Catalog No. BMA57032
Change view
Click to view available options
Separation Range:
10 to 1000 bp
50 to 4000 bp
Wells:
12 + 1-well
16 + 1-well
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Separation Range Wells
BMA57032 10 to 1000 bp 16 + 1-well
BMA57031 10 to 1000 bp 12 + 1-well
BMA57023 50 to 4000 bp 12 + 1-well
BMA57029 50 to 4000 bp 16 + 1-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BMA57032 Supplier Lonza™ Supplier No. 57032
Add to Cart
Add to Cart

Cassettes optimized for DNA migration. Lonza™ FlashGel™ System DNA Cassettes separate DNA quickly and DNA migration can be seen as it happens. Separates DNA in 2 to 7 minutes.

  • Get results in 5 minutes or less
  • Contain pre-stained agarose gels and buffer, minimizing user exposure to potentially mutagenic stains
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2.2%
Separation Range 10 to 1000 bp
Wells 16 + 1-well
Formulation 2.2% Agarose
For Use With (Application) Electrophoresis
Kit Contents Agarose Gel, Stain and Buffer
Material Agarose
Quantity 9 Pack
Storage Requirements 18° to 26°C
Length (English) Cassette 4.5 in.
Length (English) Gel 2.7 in.
Length (Metric) Cassette 11.5 cm
Length (Metric) Gel 7 cm
Width (English) Cassette 4.2 in.
Width (Metric) Cassette 10.7 cm
Width (Metric) Gel 8.4 cm
Height (English) Cassette 0.6 in.
Height (Metric) Cassette 1.7 cm
Product Type DNA Cassettes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.