Learn More
BUCHI FlashPure Chromatography Cartride Sample Packs
SDP
Catalog No. 01670195 Shop All BUCHI Products
Change view
Click to view available options
Bed Weight (Metric):
12 g, 20 g, 80 g
4 g, 12 g, 20 g, 25 g, 40 g, 80 g
Material:
C18
Silica, C18
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight (Metric) Material
01-670-195 12 g, 20 g, 80 g C18
01-670-196 4 g, 12 g, 20 g, 25 g, 40 g, 80 g Silica, C18
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 01-670-195 Supplier BUCHI Supplier No. 34004989
Add to Cart
Add to Cart
This item is not returnable. View return policy
FlashPure cartridges are offered in a wide range of sizes, covering different stationary phases, particle sizes and geometries for your purification needs.
FlashPure EcoFlex cartridges are designed with optimized backpressure, price and performance.
  • Normal Phase Bundle for basic applications and polar compounds
  • Reverse Phase C18 Bundle for nonpolar compounds and ideal for peptides
  • Scale-up Bundle for larger loading separations
Specifications
Material C18
Type Chromatography Cartridge
Includes Sample Bundle of Cartridges
Bed Weight (Metric) 12 g, 20 g, 80 g
Column Type Stationary Phase