Learn More
Description
- Most economical solution without sacrificing quality for standard separations
- Large selection of cartridge sizes from 4g-5,000g
- Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
- Silica is ideal for basic applications and polar compounds using organic solvents
- Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
- C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
- Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
- Neutral Alumina is ideal for aldehydes, ketones, and esters
Specifications
Specifications
| Material | Polypropylene |
| Type | Amino |
| Includes | 5 Pc. |
| Resolution | 40 to 60 μm |
| Size (Metric) Tube Rack | 12 g |
| Column Type | Amino |
| For Use With (Application) | Better Peak Shapes with Polar and Basic Compounds, Ideal for Carbohydrates |
| For Use With (Equipment) | Any Flash System |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
