Learn More
Chemglass Life Sciences Chemglass Erlenmeyer Flask, 250 mL, Polycarbonate, Baffled Bottom, Vent Filter Cap, Sterile
SDP
Supplier:  Chemglass Life Sciences CGN2091250
Encompass_Preferred
The Chemglass Life Sciences 250 mL polycarbonate Erlenmeyer flask is a sterile, non-pyrogenic baffled conical flask supplied with a 0.2 μm hydrophobic vent filter cap. It is designed for cell culture and bioprocess applications that require controlled gas exchange and contamination control; graduations are molded for accurate volume measurement. Flasks are sterilized by E-beam (SAL=10) and packaged individually in sterile bags.
  • Sterile, E-beam sterilized (SAL=10).
  • Made from polycarbonate resin.
  • 0.2 μm hydrophobic vent filter cap for gas exchange without contamination.
  • Quadruple baffled bottom to promote aeration and mixing.
  • Non-pyrogenic and DNase/RNase-free.
  • Individually packaged in sterile bags.
  • Molded graduations for accurate volume measurement.
  • Dimensions: height 137.8 mm; neck diameter 34 mm; bottom diameter 83 mm.
Catalog No. 50-273-3481