Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Flask Form Electroscope
SDPSCI_ED

Catalog No. S139483 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139483 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139483 Supplier United Scientific™ Supplier No. ELFL02PK/3
Add to Cart
Add to Cart
This item is not returnable. View return policy

Learn to detect and estimate charges with this device.

  • Electroscope is housed in a 250 mL flask with a metal rod attached to a rubber stopper
  • A pair of aluminum leaves is suspended from an alligator clip
  • The knob on top of the rubber stopper accepts a charge
  • When the charge enters the flask, the aluminum leaves separate

Specifications

Grade 5 to 12
Format Model
Product Type Electroscope
Includes Electroscope and Activity Guide
Languages English
Subject Physics

Warning: Cancer and reproductive harm

Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.