Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Flask Length Penny-Head Stopper
GREENER_CHOICESDP

Catalog No. 31501883 Shop All Kemtech Products
Change view
Click to view available options
Size:
13
16
19
22
27
32
38
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
31-501-883 16
31-501-882 13
31-501-884 19
31-501-885 22
31-501-886 27
31-501-887 32
31-501-888 38
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 31-501-883 Supplier Synthware™ Supplier No. S280016
Add to Cart
Add to Cart

Various Joint Sizes Available.

Specifications

For Use With (Application) Organic Synthesis
Quantity 1 Ea.
Size 16
Color Clear
Material Borosilicate Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.