Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Flask Length PFTE Stopper
GREENER_CHOICESDP

Catalog No. 31501903 Shop All Kemtech Products
Change view
Click to view available options
Stopper Number:
13
16
22
27
38
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stopper Number
31-501-903 27
31-501-900 13
31-501-901 16
31-501-902 22
31-501-904 38
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 31-501-903 Supplier Synthware™ Supplier No. S120027
Add to Cart
Add to Cart

Stopper features a PTFE handle for easy gripping.

Specifications

Stopper Number 27
For Use With (Application) Organic Synthesis
Color White
Material PTFE
Quantity 1 Ea.
Size #27
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.