Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Phoenix™ Flask Set
SCI_ED

Catalog No. S04407 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Flask set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S04407 Flask set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S04407 Supplier Aqua Phoenix™ Supplier No. GL5502SET

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for laboratory use.

  • Glass
  • Includes one each of 50, 125, 250, 500 and 1000mL

Specifications

Capacity (Metric) 50 mL, 125 mL, 150 mL, 500 mL, 1000 mL
Includes One each of 50, 125, 250, 500 and 1000mL flask
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.