Learn More
Chemglass Life Sciences 50mL Volumetric Flask, Class A, #9 Outer Stopper Neck, Glass Stopper
Supplier: Chemglass Life Sciences CG160004
50mL Volumetric Flask, Class A, #9 Outer Stopper Neck, Glass Stopper. Flasks are manufactured to conform to Federal Specifications NNN-F-289 for Type 1, Class “A” unserialized flasks and ASTM E-288. Supplied complete with one solid glass pennyhead stopper. For additional stoppers, see CG-3018.