Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ FlaskScrubber Starter Kit
GSA_VASDP

Catalog No. 09311204 Shop All Labconco Corporation Products
Change view
Click to view available options
No. of Pins:
48
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Pins
09-311-204 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-311-204 Supplier Labconco™ Supplier No. 4577200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Outfit a FlaskScrubber to wash a variety of narrow-mouth labware.

  • Use to wash a variety of narrow-mouth labware
  • Includes 24-Place Pipette Insert (4575400), Graduated Cylinder Insert (4574600), Utensil Basket with Cover (4573100), 48-Pin Insert (4574200), and Standard Upper Rack (4668700)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Material Stainless Steel
Product Type Starter Kit
Color Silver
No. of Pins 48
Rack Type Spindle
For Use With (Application) Washing Glassware
For Use With (Equipment) FlaskScrubber Glassware Washer
Includes 24-Place Pipette Insert (4575400), Graduated Cylinder Insert (4574600), Utensil Basket with Cover (4573100), 48-Pin Insert (4574200), and Standard Upper Rack (4668700)
Dimensions (L x W x H) 17.7 x 8.9 x 5.1 in.
Weight (English) Shipping 20 lb.
Weight (Metric) Shipping 9.1 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.