Learn More
Opentrons Labworks Inc Universal Flat Adapter for Heater-Shaker Module
Supplier:  Opentrons Labworks Inc 99100067
Encompass
Thermal adapter for a range of ANSI/SLAS compliant labware, e.g., Corning 384 Well Plate 112 µL. Compatible with the Opentrons Heater-Shaker Module.
Catalog No. NC2752932
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.